missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP75 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14887-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognized the endogenous HSPA9 protein in NIH/3T3 cells.
The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.Specifications
GRP75 | |
Polyclonal | |
Unconjugated | |
HSPA9 | |
CSA, GRP 75, GRP75, Heat shock 70 kDa protein 9, HSPA9, HSPA9B, Mortalin, MOT, mot 2, MOT2, MTHSP75, PBP74, Peptide binding protein 74 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15526, 291671, 3313 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P38646, P38647, P48721 | |
HSPA9 | |
GRP75 Fusion Protein Ag6673 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |