missing translation for 'onlineSavingsMsg'
Learn More

GRWD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10354-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826693

  • 1236.20 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GRWD1 is a component of the 50S and 80S preribosomal complexes and plays a role in ribosome biogenesis (Gratenstein et al., 2005).
Specifications

Specifications

GRWD1
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5XI13, Q810D6, Q9BQ67
GRWD, GRWD1, KIAA1942, RRB1, WDR28
Rabbit
IgG
20 μL
-20°C
Primary
101612, 308592, 83743
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GRWD1
Liquid
Grwd1
GRWD1 Fusion Protein Ag0367
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers