Läs mer
Tocris Bioscience™ GS 143
Beskrivning
GS 143 is a β-TrCP1 ligase inhibitor. Inhibits IκBα ubiquitination (IC50 = 5.2 μM) without affecting proteasome activity. Blocks LPS-induced expression of inflammatory cytokines in human myelomonocytic cells. Suppresses antigen-induced NFκB expression, inflammation and mucus production in airways of OVA-sensitized mice.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-[4-[[5-(2-Fluorophenyl)-2-furanyl]methylene]-4,5-dihydro-5-oxo-3-(phenylmethyl)-1H-pyrazol-1-yl]benzoic acid |
| CAS | 916232-21-8 |
| Kvantitet | 10 mg |
| Mål | Ubiquitin Ligase (E3) Inhibitors |
| Molekylformel | C28H19FN2O4 |
| Formel vikt | 466.46 |
| Renhet | >98% |