Läs mer
- Home
- Products
- Personlig skyddsutrustning
- Handskydd
- Torra boxhandskar
- AL Glove Systems GSB Isolator Box Gloves
AL Glove Systems GSB Isolator Box Gloves
Produktkod. 31337494| Produktkod. | Storlek | Längd (metrisk) | Tjocklek (metrisk) | Diameter (metrisk) manschett |
|---|---|---|---|---|
| 31337494 | Large, 9.5 | 920 mm | 0.4 mm | 250 mm |
| 31337400 | Medium, 8 | 800 mm | 0.4 mm | 180 mm |
| 31337436 | Medium, 8 | 800 mm | 0.6 mm | 180 mm |
| 31337484 | Medium, 8 | 800 mm | 0.4 mm | 200 mm |
| 31337398 | Medium, 8 | 800 mm | 0.6 mm | 200 mm |
| 31337454 | Medium, 8 | 800 mm | 0.4 mm | 225 mm |
| 31337483 | Medium, 8 | 800 mm | 0.6 mm | 225 mm |
| 31337482 | Large, 9.5 | 800 mm | 0.4 mm | 160 mm |
| 31337497 | Large, 9.5 | 800 mm | 0.6 mm | 160 mm |
| 31337429 | Large, 9.5 | 800 mm | 0.4 mm | 180 mm |
| 31337439 | Large, 9.5 | 800 mm | 0.6 mm | 200 mm |
| 31337416 | Large, 9.5 | 800 mm | 0.4 mm | 225 mm |
| 31337517 | Large, 9.5 | 800 mm | 0.4 mm | 250 mm |
| 31337495 | Large, 9.5 | 800 mm | 0.6 mm | 250 mm |
| 31337424 | Large, 9.5 | 800 mm | 0.4 mm | 300 mm |
| 31337508 | Large, 9.5 | 800 mm | 0.6 mm | 300 mm |
| 31337461 | Large, 9.5 | 920 mm | 0.4 mm | 200 mm |
| 31337476 | Large, 9.5 | 920 mm | 0.6 mm | 200 mm |
| 31337469 | Large, 9.5 | 920 mm | 0.6 mm | 250 mm |
| 31337396 | X-Large, 11 | 800 mm | 0.6 mm | 200 mm |
| 31337407 | X-Large, 11 | 800 mm | 0.4 mm | 225 mm |
| 31337493 | X-Large, 11 | 800 mm | 0.4 mm | 250 mm |
| 31337419 | X-Large, 11 | 800 mm | 0.6 mm | 250 mm |
| 31337401 | X-Large, 11 | 800 mm | 0.4 mm | 300 mm |
| 31337457 | X-Large, 11 | 920 mm | 0.4 mm | 300 mm |
| 31337518 | X-Large, 11 | 800 mm | 0.6 mm | 300 mm |
| 31337399 | X-Large, 11 | 920 mm | 0.4 mm | 225 mm |
| 31337467 | X-Large, 11 | 920 mm | 0.6 mm | 225 mm |
| 31337447 | X-Large, 11 | 920 mm | 0.4 mm | 250 mm |
| 31337488 | X-Large, 11 | 920 mm | 0.6 mm | 250 mm |
| 31337462 | X-Large, 11 | 920 mm | 0.6 mm | 300 mm |
Beschreibung
AL Glove Systems GSB gloves are made of bromo-butyl rubber (BBR) with high gas impermeability and high resistance to a wide range of polar hydrocarbons such as acids, esters, ketones, and amine derivatives. They are highly flexible with a high sensitivity. The GSB Glove is classified as PAH-free.
- Impermeable to gas
- High resistance to the permeation of mainly oxygen, nitrogen, inert gases, etc. during production under inert gas atmosphere in inert boxes for the production of electronic components and microcomponents
Flexible:
- Offers good sensitivity in handling
- Resistant to chemicals
- High resistance to a wide range of chemicals and toxins
PERMEATION EN ISO 374-1:2016/A1:2018
- A Methanol Protection Level 6
- B Acetone Protection Level 6
- I Ethyl acetate Protection Level 3
- K Sodium hydroxide 40% Protection Level 6
- L Sulphuric acid 96% Protection Level 6
- N Acetic acid 99% Protection Level 6
- O Ammonium hydroxide 25% Protection Level 6
- T Formaldehyde 37% Protection Level 6
Spezifikation
Spezifikation
| Certifieringar/efterlevnad (Europa) | PPE Category III Directive 2016/425 EC Type |
| Klorerad | No |
| Färg | Black |
| Hand | Ambidextrous |
| Latexfri | Yes |
| Material | Butyl |
| Produkttyp | Dry Box, Isolator Gloves |
| Storlek | Large, 9.5 |
| Temperatur (engelska) Max. | 194°F |
| Temperatur (metrisk) Max | 90°C |
| Tjocklek (engelska) | 16 mil |
| Tjocklek (metrisk) | 0.4 mm |
| För användning med (applikation) | Handling Sensitive or Hazardous Materials in an Inert or Contained Atmosphere |
| För användning med (utrustning) | Dry Box, Isolator, RABS |
| Greppdesign | Smooth |
| Tillverkning/QMS revisionsstandarder | ISO 9001:2015 |
| Förpackningstyp | Carton |
| Temperatur (engelska) Min | -40°F |
| Temperatur (metrisk) Min. | -40°C |
| Längd (engelska) | 36 in. |
| Längd (metrisk) | 920 mm |
| Diameter (engelska) manschett | 10 In. |
| Diameter (metrisk) manschett | 250 mm |
| Mehr anzeigen Weniger anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.