Läs mer
GSK 1016790A, TRC
Produktkod. 30240614
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240614 10 mg
30240784 25 mg
30240917 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240614 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG79744010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 942206-85-1
Molekylformel C28H32Cl2N4O6S2
Molekylvikt (g/mol) 655.61
Synonym N-[(1S)-1-[[4-[(2S)-2-[[(2,4-dichlorophenyl)sulfonyl]amino]-3-hydroxy-1-oxopropyl]-1-piperazinyl]carbonyl]-3-methylbutyl]benzo[b]thiophene-2-carboxamide, N-((S)-1-(4-((S)-2-(2,4-Dichlorophenylsulfonamido)-3-hydroxypropanoyl)piperazin-1-yl)-4-methyl-1-oxopentan-2-yl)benzo[b]thiophene-2-carboxamide
IUPAC-namn N-[(2S)-1-[4-[2-[(2,4-dichlorophenyl)sulfonylamino]-3-hydroxypropanoyl]piperazin-1-yl]-4-methyl-1-oxopentan-2-yl]-1-benzothiophene-2-carboxamide
LEDER O=C(N[C@@H](CC(C)C)C(N1CCN(C(C(NS(C2=C(Cl)C=C(Cl)C=C2)(=O)=O)CO)=O)CC1)=O)C3=CC4=C(C=CC=C4)S3
Specifikationer
Kemiskt namn eller material GSK 1016790A
InChI formel InChI=1S/C28H32Cl2N4O6S2/c1-17(2)13-21(31-26(36)24-14-18-5-3-4-6-23(18)41-24)27(37)33-9-11-34(12-10-33)28(38)22(16-35)32-42(39,40)25-8-7-19(29)15-20(25)30/h3-8,14-15,17,21-22,32,35H,9-13,16H2,1-2H3,(H,31,36)/t21-,22?/m0/s1
Kvantitet 10 mg
Formel vikt 654.114