Läs mer
Tocris Bioscience™ GSK 2033
Potent LXR antagonist
Brand: Tocris Bioscience™ 5694/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
GSK 2033 is a potent LXR antagonist (pIC50 = 7.5). Enhances T-cell proliferation and blocks T 0901317 -antiproliferative activity on T-cells. Cell permeable.Specifikationer
| 2,4,6-Trimethyl-N-[[3′-(methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl]-N-[[5-(trifluoromethyl)-2-furanyl]methyl]benzenesulfonamide | |
| 1221277-90-2 | |
| 50 mg |