Läs mer
Tocris Bioscience™ GSK 2334470
Produktkod. 18095642
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18095642 10 mg
18064462 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18095642 Leverantör Tocris Bioscience™ Leverantörsnummer 4143/10
Highly potent, selective PDPK1 (PDK1) inhibitor
Potent 3-phosphoinositide-dependent protein kinase (PDPK1) inhibitor (IC50 ∽ 10 nM). Exhibits no effect on other kinases including Aurora, ROCK, p38 MAPK and PI 3-K. Suppresses T-loop phosphorylation and subsequent activation of PDK1 substrates S6K1, SGK and RSK in vitro; exhibits limited inhibitory effect on Akt activation. Delays melanomagenesis and metastasis in BrafV600E::Pten(-/-) mice.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1227911-45-6
Molekylformel C25H34N8O
Specifikationer
Kemiskt namn eller material GSK 2334470
Kvantitet 10 mg
Formel vikt 462.59
Renhet >98%
Molekylvikt (g/mol) 462.602