Läs mer
Tocris Bioscience™ GSK 319347A
Potent and selective IKKε inhibitor
Brand: Tocris Bioscience™ 5672/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
GSK 319347A is a potent and selective IKKε inhibitor (pIC50 = 7.4). Exhibits >100-fold selectivity for IKKε over IKKα IKKβ, and a range of other kinases.Specifikationer
| Potent and Selective Inhibitor | |
| 862812-98-4 | |
| 469.53g/mol | |
| IkB Kinase Inhibitors |
| IkB Kinase | |
| C22H19N3O5S2 | |
| Store at -20°C | |
| 10 mg |
Produkttitel
For Research Use Only