Läs mer
Tocris Bioscience™ GSK J1
GREENER_CHOICE
Produktkod. 18055332
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18055332 50 mg
18034352 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18055332 Leverantör Tocris Bioscience™ Leverantörsnummer 4593/50
Potent JMJD3/UTX inhibitor
Potent inhibitor of the H3K27 histone demethylases JMJD3 (KDM6B) and UTX (KDM6A) (IC50 values are 28 and 53 nM respectively). Also inhibits KDM5B, KDM5C and KDM5A (IC50 values are 170, 550 and 6,800 nM respectively). Exhibits no activity against a panel of other histone demethylases (IC50 >20 μM), and displays no significant inhibitory activity against 100 protein kinases at a concentration of 30 μM.
Ethyl ester derivative and Negative Control also available.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1373422-53-7
Molekylformel C22H23N5O2
Specifikationer
Kemiskt namn eller material GSK J1
Kvantitet 50 mg
Formel vikt 389.45
Renhet >99%
Molekylvikt (g/mol) 389.459