Läs mer
GSK-J4, MedChemExpress
Produktkod. 16472797
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
Förpackningsstorlek:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16472797 50 mg 50mg
16456718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16472797 Leverantör Medchem Express Leverantörsnummer HY15648B
MedChemExpress GSK-J4 is a potent dual inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A with IC50s of 8.6 and 6.6 μM, respectively. GSK-J4 inhibits LPS-induced TNF-α production in human primary macrophages with an IC50 of 9 μM. GSK J4 is a cell permeable prodrug of GSK-J1. GSK-J4 induces endoplasmic reticulum stress-related apoptosis.
GSK-J4 is a potent dual inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A with IC50s of 8.6 and 6.6 μM, respectively. GSK-J4 inhibits LPS-induced TNF-α production in human primary macrophages with an IC50 of 9 μM. GSK J4 is a cell permeable prodrug of GSK-J1. GSK-J4 induces endoplasmic reticulum stress-related apoptosis.
Specifikationer
Kemiskt namn eller material GSK-J4
Fysisk form Oil
CAS 1373423-53-0
Färg Yellow
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C24H27N5O2
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : ≥ 36 mg/mL (86.23 mM)
LEDER O=C(OCC)CCNC1=NC(C2=CC=CC=N2)=NC(N3CCC(C=CC=C4)=C4CC3)=C1
Molekylvikt (g/mol) 417.5
Formel vikt 417.5
Procent renhet 95.0%