Läs mer
GSK PERK Inhibitor, TRC
Produktkod. 30179483
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179483 5 mg
30179160 1 mg
30179479 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179483 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG7978005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1337531-89-1
Molekylformel C24 H19 F4 N5 O
Molekylvikt (g/mol) 469.43
Synonym GSK PERK Inhibitor, 1-[5-(4-Amino-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-5-yl)-2,3-dihydro-1H-indol-1-yl]-2-[3-fluoro-5-(trifluoromethyl)phenyl]ethanone
IUPAC-namn 1-[5-(4-amino-7-methyl-pyrrolo[2,3-d]pyrimidin-5-yl)indolin-1-yl]-2-[3-fluoro-5-(trifluoromethyl)phenyl]ethanone
LEDER Cn1cc(c2ccc3N(CCc3c2)C(=O)Cc4cc(F)cc(c4)C(F)(F)F)c5c(N)ncnc15
Specifikationer
Kemiskt namn eller material 3-Fluoro-GSK2606414
Kvantitet 5 mg
InChI formel InChI=1S/C24H19F4N5O/c1-32-11-18(21-22(29)30-12-31-23(21)32)14-2-3-19-15(9-14)4-5-33(19)20(34)8-13-6-16(24(26,27)28)10-17(25)7-13/h2-3,6-7,9-12H,4-5,8H2,1H3,(H2,29,30,31)
Formel vikt 469.1526