Läs mer
GSK126, MedChemExpress
Produktkod. 16478397
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
200 mg
Förpackningsstorlek:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16478397 5 mg -
16466196 100 mg 100mg
16461867 50 mg -
16445708 10 mg -
16431086 200 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16478397 Leverantör Medchem Express Leverantörsnummer HY13470
MedChemExpress GSK126 (GSK2816126A) is a potent, highly selective inhibitor of EZH2 methyltransferase with an IC50 of 9.9 nM.
GSK126 (GSK2816126A) is a potent, highly selective inhibitor of EZH2 methyltransferase with an IC50 of 9.9 nM.
Specifikationer
Kemiskt namn eller material GSK126
CAS 1346574-57-9
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C31H38N6O2
Kvantitet 5 mg
Synonym GSK2816126A
Löslighetsinformation DMSO : 12.5 mg/mL (23.73 mM; Need ultrasonic)
LEDER O=C(C1=CC(C2=CC=C(N3CCNCC3)N=C2)=CC4=C1C(C)=CN4[C@@H](C)CC)NCC5=C(C)C=C(C)NC5=O
Molekylvikt (g/mol) 526.67
Formel vikt 526.67
Procent renhet 95.0%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-programmed cell death