missing translation for 'onlineSavingsMsg'
Läs mer

GSK2837808A, MedChemExpress

Product Code. 16485847
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
16485847 5 mg -
16463118 10 mg 10mg
16429676 50 mg -
16412636 25 mg -
16410036 2 mg -
16404416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16485847 Supplier Medchem Express Supplier No. HY100681

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2837808A is a potent and selective lactate dehydrogenase A (LDHA) inhibitor with IC50s of 2.6 and 43 nM for hLDHA and hLDHB, respectively.

GSK2837808A is a potent and selective lactate dehydrogenase A (LDHA) inhibitor with IC50s of 2.6 and 43 nM for hLDHA and hLDHB, respectively.

Specifications

Kemiskt namn eller material GSK2837808A
CAS 1445879-21-9
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C31H25F2N5O7S
Kvantitet 5 mg
Löslighetsinformation DMSO : 50 mg/mL (76.97 mM; Need ultrasonic)
LEDER O=C(O)C1=CC(OC2=CC(F)=CC(F)=C2)=CC(NC3=C(S(=O)(NC4CC4)=O)C=NC5=CC(C6=CN=C(OC)N=C6OC)=CC=C35)=C1
Molekylvikt (g/mol) 649.62
Formel vikt 649.62
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.