Läs mer
GSK2982772, MedChemExpress
Produktkod. 16476277
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16476277 5 mg -
16469966 50 mg -
16460899 1 mg -
16444666 100 mg -
16443558 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16476277 Leverantör Medchem Express Leverantörsnummer HY101760
MedChemExpress GSK2982772 is a potent, orally active and ATP competitive RIP1 kinase inhibitor with IC50 values of 16 nM and 20 nM for human and monkey RIP1, respectively.
GSK2982772 is a potent, orally active and ATP competitive RIP1 kinase inhibitor with IC50 values of 16 nM and 20 nM for human and monkey RIP1, respectively.
Specifikationer
Kemiskt namn eller material GSK2982772
CAS 1622848-92-3
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C20H19N5O3
Kvantitet 5 mg
Löslighetsinformation DMSO : 250 mg/mL (662.43 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
LEDER O=C(C1=NC(CC2=CC=CC=C2)=NN1)N[C@H]3COC4=CC=CC=C4N(C)C3=O
Molekylvikt (g/mol) 377.4
Formel vikt 377.4
Procent renhet 95.23%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation