missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GSK3B (Human) Recombinant Protein
Human GSK3B full-length ORF ( AAH00251, 1 a.a. - 433 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002932-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSGRPRTTSFAESCKPVQQPSAFGSMKVSRDKDGSKVTTVVATPGQGPDRPQEVSYTDTKVIGNGSFGVVYQAKLCDSGELVAIKKVLQDKRFKNRELQIMRKLDHCNIVRLRYFFYSSGEKKDEVYLNLVLDYVPETVYRVARHYSRAKQTLPVIYVKLYMYQLFRSLAYIHSFGICHRDIKPQNLLLDPDTAVLKLCDFGSAKQLVRGEPNVSYICSRYYRAPELIFGATDYTSSIDVWSAGCVLAELLLGQPIFPGDSGVDQLVEIIKVLGTPTREQIREMNPNYTEFKFPQIKAHPWTKDSSGTGHFTSGVRVFRPRTPPEAIALCSRLLEYTPTARLTPLEACAHSFFDELRDPNVKLPNGRDTPALLNFTTQELSSNPPLATILIPPHARIQAAASTPTNATAASDANTGDRGQTNNAASASASNST
Specifications
AAH00251 | |
glycogen synthase kinase 3 beta | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2932 | |
Wheat germ expression system | |
10 μg | |
GSK3B | |
GST |