Learn More
GSK3B Mouse anti-Human, Unconjugated, Clone: 2E9E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67329-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The GSK3B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSK3B. This antibody recognizes human, pig antigen. The GSK3B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
GSK3B | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA | |
2E9E4 | |
Unconjugated | |
150 μL | |
2932 | |
IgG1 |