Läs mer
GSK682753A, MedChemExpress
Produktkod. 16486017
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16486017 5 mg -
16492706 25 mg 25mg
16453298 10 mg -
16449836 50 mg -
16464546 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16486017 Leverantör Medchem Express Leverantörsnummer HY101192
MedChemExpress GSK682753A is a selective and highly potent inverse agonist of the epstein-barr virus-induced receptor 2 (EBI2) with an IC50 of 53.6 nM.
GSK682753A is a selective and highly potent inverse agonist of the epstein-barr virus-induced receptor 2 (EBI2) with an IC50 of 53.6 nM.
Specifikationer
Kemiskt namn eller material GSK682753A
Fysisk form Powder
CAS 1334294-76-6
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C23H21Cl3N2O3
För användning med (applikation) COVID-19-anti-virus
Löslighetsinformation DMSO : ≥ 100 mg/mL (208.43 mM)
LEDER O=C1OC2(CCN(C(/C=C/C3=CC=C(Cl)C=C3)=O)CC2)CN1CC4=CC=C(Cl)C(Cl)=C4
Molekylvikt (g/mol) 479.78
Formel vikt 479.78
Procent renhet 99.56%