missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSS Mouse anti-Human, Unconjugated, Clone: 1F12G1, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GSS |
---|---|
Clone | 1F12G1 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840316
|
Proteintech
67598-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830316
|
Proteintech
67598-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The GSS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSS. This antibody recognizes human, mouse, rat, pig antigen. The GSS antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
GSS | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
2937 | |
Primary |
1F12G1 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG1 |