Learn More
GSTT1 Mouse anti-Human, Unconjugated, Clone: 1A4C11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67645-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16870386
  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.
Description
Description
The GSTT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSTT1. This antibody recognizes human antigen. The GSTT1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GSTT1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1A4C11
Unconjugated
150 μL
2952
IgG2b