missing translation for 'onlineSavingsMsg'
Learn More

GSTZ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1236.20 SEK - 3313.85 SEK

Specifications

Antigen GSTZ1
Concentration 0.13 mg/mL
Applications Western Blot
Conjugate Unconjugated
Format Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16813784
View Documents Promotion Details
Proteintech
14889-1-AP-150UL
150 μL
3313.85 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 20-01-2022
Please to purchase this item. Need a web account? Register with us today!
16823784
View Documents Promotion Details
Proteintech
14889-1-AP-20UL
20 μL
1236.20 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 20-01-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene is a member of the glutathione S-transferase (GSTs) super-family which encodes multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Thus defects in this enzyme may lead to severe metabolic disorders including alkaptonuria, phenylketonuria and tyrosinaemia. Several transcript variants of this gene encode multiple protein isoforms.
Specifications

Specifications

GSTZ1
Western Blot
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Polyclonal
-20°C
0.13 mg/mL
Unconjugated
Rabbit
Rat, Mouse, Human
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications