missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTDC1 Mouse anti-Human, Porcine, Clone: 1C4C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66578-1-IG-20UL
Additional Details : Weight : 0.00010kg
Specifications
GTDC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4AE62 | |
GTDC1 | |
GTDC1 Fusion Protein Ag9934 | |
20 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1C4C10 | |
Unconjugated | |
GTDC1 | |
GTDC1, mat Xa | |
Mouse | |
Protein G | |
RUO | |
100152895, 79712 | |
-20°C | |
Liquid |