missing translation for 'onlineSavingsMsg'
Learn More

GTF2A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10540-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16896983

  • 1656.73 SEK / 20µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A), TFIID TFIIE TFIIF TFIIG/TFIIJ, and TFIIH respectively, and a 12-kD gamma subunit encoded by GTF2A2.
Specifications

Specifications

GTF2A2
Polyclonal
Unconjugated
GTF2A2
GTF2A2, HsT18745, TF2A2, TFIIA, TFIIA 12, TFIIA gamma, TFIIA p12 subunit, TFIIAS
Rabbit
Antigen Affinity Chromatography
RUO
235459, 2958, 83828
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08950, P52657, Q80ZM7
Gtf2a2
GTF2A2 Fusion Protein Ag0825
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers