missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ GTF2E2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582166
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: LLEDIEEALP NSQKAVKALG DQILFVNRPD KKKILFFNDK SCQFSVDEEF QKLWRSVTVD SMDEEKIEEY LKRQGISSMQ ESGPKKVAPI QRRKKPASQK KRRFKTHNEH LAGVLKDYSD ITSSK Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
Specifikationer
| GTF2E2 | |
| Polyclonal | |
| Unconjugated | |
| GTF2E2 | |
| 34kDa; AI462509; C330006J08Rik; FE; general transcription factor II E, polypeptide 2 (beta subunit); general transcription factor II E, polypeptide 2 (beta subunit, 34kD); general transcription factor IIE subunit 2; general transcription factor IIE, polypeptide 2 (beta subunit); general transcription factor IIE, polypeptide 2 (beta subunit, 34kD); general transcription factor IIE, polypeptide 2, beta; general transcription factor IIE, polypeptide 2, beta 34kDa; Gtf2e2; T2EB; TF2E2; TFIIE beta subunit; TFIIE-B; TFIIE-beta; transcription initiation factor IIE subunit beta; TTD6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2961 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P29084 | |
| GTF2E2 | |
| Recombinant protein corresponding to Human GTF2E2. Recombinant protein control fragment (Product #RP-104547). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering