Learn More
GTPBP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565097
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: EFQVDEIYTVP EVGTVVGGTL SSGICREGDQ LVVGPTDDGC FLELRVCSIQ RNRSACRVLR AGQAATLALG DFDRALLRKG MVMVSPEM Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
GTPBP2 Polyclonal antibody specifically detects GTPBP2 in Human samples. It is validated for Immunocytochemistry.Specifications
GTPBP2 | |
Polyclonal | |
Unconjugated | |
Gtpbp2 | |
GTP binding protein 2; GTP-binding protein 2; GTP-binding-like protein 2; gtpbp2; I79_024595; nmf205 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BX10 | |
Gtpbp2 | |
Recombinant Human GTPBP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only