Läs mer
Invitrogen™ GTPBP6 Polyclonal Antibody
Beskrivning
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PRDQLFATLD VTAHAGTLPS RMTVLYVDTI GFLSQLPHGL IESFSATLED VAHSDLILHV RDVSHPEAEL QKCSVLSTLR GLQLPAPLLD SMVEVHNKVD LVPGYSPTEP NVVPVS Highest antigen sequence indentity to the following orthologs: Rat - 71%, Mouse - 72%.
Specifikationer
Specifikationer
| Antigen | GTPBP6 |
| Användningsområden | Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 0.1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gen | GTPBP6 |
| Genaccessionsnr. | O43824 |
| Gene Alias | AV119224; GTP binding protein 6 (putative); Gtpbp6; Pgbpll; pgpl; pseudoautosomal GTP binding protein-like, like; Pseudoautosomal GTP-binding protein-like; Putative GTP-binding protein 6; RGD1305954 |
| Gensymboler | GTPBP6 |
| Visa mer |