Läs mer
Guaiol, TRC
Produktkod. 17937334
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17937334 50 mg
17937904 10 mg
17958974 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17937334 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG80550550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 489-86-1
Molekylformel C15 H26 O
Molekylvikt (g/mol) 222.37
Synonym 5-Azulenemethanol, 1,2,3,4,5,6,7,8-octahydro-α,α,3,8-tetramethyl-, (3S,5R,8S)-, 5-Azulenemethanol, 1,2,3,4,5,6,7,8-octahydro-α,α,3,8-tetramethyl-, [3S-(3α,5α,8α)]-, 5-Azulenemethanol, 1,2,3,4,5β,6,7,8-octahydro-α,α,3α,8α-tetramethyl- (7CI), Guai-1(5)-en-11-ol (8CI), Guaiol (6CI), (3S,5R,8S)-1,2,3,4,5,6,7,8-Octahydro-α,α,3,8-tetramethyl-5-azulenemethanol, (-)-Guaiol, 3,8-Dimethyl-5-α-hydroxyisopropyl-δ9-octahydroazulene, Champaca camphor, Champacol, Guaiac alcohol, NSC 19941
IUPAC-namn 2-[(3S,5R,8S)-3,8-dimethyl-1,2,3,4,5,6,7,8-octahydroazulen-5-yl]propan-2-ol
LEDER C[C@H]1CC[C@H](CC2=C1CC[C@@H]2C)C(C)(C)O
Specifikationer
Kemiskt namn eller material Guaiol
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C15H26O/c1-10-5-7-12(15(3,4)16)9-14-11(2)6-8-13(10)14/h10-12,16H,5-9H2,1-4H3/t10-,11-,12+/m0/s1
Formel vikt 222.1984
Procent renhet >95%