Läs mer
Guanabenz Acetate, MedChemExpress
Produktkod. 16498366
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16498366 100 mg
16464397 50 mg
16432176 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16498366 Leverantör Medchem Express Leverantörsnummer HYB0566
MedChemExpress Guanabenz (Acetate) (BR-750) is an alpha-2 selective adrenergic agonist used as an antihypertensive agent.
Guanabenz (Acetate) (BR-750) is an alpha-2 selective adrenergic agonist used as an antihypertensive agent.
Specifikationer
Kemiskt namn eller material Guanabenz Acetate
Fysisk form Solid
CAS 23256-50-0
Färg Light Yellow
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C10H12Cl2N4O2
För användning med (applikation) COVID-19-immunoregulation
Synonym BR-750 Wy8678 acetate
Löslighetsinformation DMSO : ≥ 51 mg/mL (175.18 mM)
LEDER ClC1=C(/C=N/NC(N)=N)C(Cl)=CC=C1.CC(O)=O
Molekylvikt (g/mol) 291.13
Formel vikt 291.13
Procent renhet 98.39%