Läs mer
Guanfacine-13C, 15N3, TRC
Produktkod. 30180374
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180374 10 mg
30179915 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180374 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG81600210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1189924-28-4
Molekylformel 13C C8 H9 Cl2 15N3 O
Molekylvikt (g/mol) 250.07
Synonym Benzeneacetamide-15N, N-(amino-15N-imino-15N-methyl-13C)-2,6-dichloro- (ACI), N-(Amino-15N-imino-15N-methyl-13C)-2,6-dichlorobenzeneacetamide-15N (ACI), [13C,15N3]Guanfacine, Guanfacine-13C,15N3, Guanfacine-13C,15N3 (guanidine-13C,15N3), [(2,6-Dichlorophenyl)acetyl]guanidine-13C-15N3, N-(Amidino-13C-15N2)-2-(2,6-dichlorophenyl)acetamide-15N
IUPAC-namn N-(C-(^{15}N)azanylcarbonimidoyl)-2-(2,6-dichlorophenyl)acetamide
LEDER [15NH2][13C](=[15NH])[15NH]C(=O)Cc1c(Cl)cccc1Cl
Specifikationer
Kemiskt namn eller material Guanfacine-13C, 15N3
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C9H9Cl2N3O/c10-6-2-1-3-7(11)5(6)4-8(15)14-9(12)13/h1-3H,4H2,(H4,12,13,14,15)/i9+1,12+1,13+1,14+1
Formel vikt 249.0067
Procent renhet >95%