missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Hydrochloride, 99+%, MP Biomedicals
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
3 kg
Specifications
Assay Percent Range | 99+% |
Quantity | 25 g |
Formula Weight | 95.53 |
Chemical Name or Material | Guanidine Hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'