Läs mer
Guanidinoacetic-13C2 Acid, TRC
Produktkod. 30005704
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005704 1 mg
30008362 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005704 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG8212521MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 634616-40-3
Molekylformel 13C2 C H7 N3 O2
Molekylvikt (g/mol) 119.09
Synonym Glycine-13C2, N-(aminoiminomethyl)- (9CI), N-(Aminoiminomethyl)glycine-1,2-13C2, Guanidineacetic-1,2-13C2 Acid, Guanidinoacetic Acid-1,2-13C2, 2-Guanidinoacetic-1,2-13C2 acid, Guanidinoacetic-13C2 Acid
IUPAC-namn 2-carbamimidamidoacetic acid
LEDER NC(=N)N[13CH2][13C](=O)O
Specifikationer
Kemiskt namn eller material Guanidinoacetic-13C2 Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C3H7N3O2/c4-3(5)6-1-2(7)8/h1H2,(H,7,8)(H4,4,5,6)/i1+1,2+1
Formel vikt 119.0605
Procent renhet >95%