Läs mer
Guanidinoacetic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Glycocyamine (Guanidinoacetic acid) |
| CAS | 352-97-6 |
| Molekylformel | C3 H7 N3 O2 |
| Kvantitet | 25 g |
| Synonym | Glycine, N-amidino- (8CI), N-(Aminoiminomethyl)glycine, 2-(Diaminomethylideneazaniumyl)acetate, 2-Carbamimidamidoacetic acid, 2-Guanidinoacetic acid, 2-[(Diaminomethylidene)amino]acetic acid, 2-[[Amino(imino)methyl]amino]acetic acid, Acetic acid, [(aminoiminomethyl)amino]-, Betacyamine, Betasyamine, Glycocyamine, Guanidine, (carboxymethyl)-, Guanidineacetic acid, Guanidinoacetic acid, Guanidoacetic acid, Guanidylacetic acid, Guanyl glycine, N-Amidinoglycine, NSC 1901, NSC 227847, NSC 26360, α-Guanidinoacetic acid, β-Guanidinoacetic acid, Glycine, N-(aminoiminomethyl)- |
| InChI formel | InChI=1S/C3H7N3O2/c4-3(5)6-1-2(7)8/h1H2,(H,7,8)(H4,4,5,6) |
| LEDER | NC(=N)NCC(=O)O |
| IUPAC-namn | 2-carbamimidamidoacetic acid |
| Molekylvikt (g/mol) | 117.11 |
| Formel vikt | 117.0538 |