Läs mer
Guanine deaminase Antibody, Novus Biologicals™
Produktkod. 18737393 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Förpackningsstorlek:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. unitSize
18737393 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 18737393 Leverantör Novus Biologicals Leverantörsnummer NBP187525
Rabbit Polyclonal Antibody
Guanine deaminase Polyclonal specifically detects Guanine deaminase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifikationer
Klassificering Polyclonal
Innehåll och lagring Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.