Läs mer
Guanosine-13C,15N2 Hydrate, TRC
Produktkod. 30006323
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006323 10 mg
30009923 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006323 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG83790210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 197227-95-5
Molekylformel 13C C9 H13 15N2 N3 O5 . H2 O
Molekylvikt (g/mol) 304.24
Synonym 2-Amino-1,9-dihydro-9-β-D-ribofuranosyl-6H-purin-6-one-13C,15N2, DL-Guanosine-13C,15N2, Guanine Ribonucleoside-13C,15N2, 9-β-D-Ribofuranosyl-
IUPAC-namn 2-(^{15}N)azanyl-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one;hydrate
LEDER O.[15NH2][13C]1=Nc2c(ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)[15NH]1
Specifikationer
Kemiskt namn eller material Guanosine-13C,15N2 hydrate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C10H13N5O5.H2O/c11-10-13-7-4(8(19)14-10)12-2-15(7)9-6(18)5(17)3(1-16)20-9;/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19);1H2/t3-,5-,6-,9-;/m1./s1/i10+1,11+1,14+1;
Formel vikt 304.0997
Procent renhet >95%