Läs mer
Guanosine 3’,5’-Cyclic Monophosphate Sodium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Guanosine 3',5'-Cyclic Monophosphate Sodium Salt |
| CAS | 40732-48-7 |
| Molekylformel | C10 H11 N5 O7 P . Na |
| Kvantitet | 10 mg |
| Synonym | Guanosine, cyclic 3',5'-(hydrogen phosphate), sodium salt (1:1), Guanosine, cyclic 3',5'-(hydrogen phosphate), monosodium salt (9CI), 4H-Furo[3,2-d]-1,3,2-dioxaphosphorin, guanosine deriv., Cyclic 3',5'-guanosine monophosphate monosodium salt, Guanosine monophosphate monosodium salt, Monosodium-GMP, Guanosine 3',5'-cyclic monophosphate sodium salt |
| InChI formel | InChI=1S/C10H12N5O7P.Na/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(21-9)1-20-23(18,19)22-6;/h2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17);/q;+1/p-1/t3-,5-,6-,9-;/m1./s1 |
| LEDER | [Na+].NC1=Nc2c(ncn2[C@@H]3O[C@@H]4COP(=O)([O-])O[C@H]4[C@H]3O)C(=O)N1 |
| IUPAC-namn | sodium;9-[(4aR,6R,7R,7aS)-7-hydroxy-2-oxido-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-2-amino-1H-purin-6-one |
| Molekylvikt (g/mol) | 367.19 |
| Formel vikt | 367.0294 |