Läs mer
MedChemExpress Guanosine 5'-triphosphate trisodium salt hydrate
Produktkod. 16411507
Encompass_Preferred
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
16411507 50 mg
1 options
Produktkod. 16411507 Leverantör MedChemExpress Leverantörsnummer HY12695B
MedChemExpress 5'-GTP trisodium salt hydrate is an activator of the signal transducing G proteins and also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.
Specifikationer
Produkttyp Reference compound
Innehåll och lagring -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Frakt skick Room temperature
Kvantitet 50 mg
Löslighetsinformation H2O : ≥ 150 mg/mL
CAS 207300-85-4
Synonym 5'-GTP trisodium salt hydrate