Läs mer
Guanylmelamine Dihydrochloride, TRC
Produktkod. 30240925
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240925 10 mg
30240478 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240925 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG41622510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27369-25-1
Molekylformel C4H8N8 • 2HCl
Molekylvikt (g/mol) 241.08
Synonym N-(4-amino-3,6-dihydro-6-imino-1,3,5-triazin-2-yl)guanidine Dihydrochloride, (4,6-diamino-1,3,5-triazin-2-yl)guanidine Dihydrochloride, (4,6-Diamino-s-triazin-2-yl)guanidine, Dihydrochloride (Aminoiminomethyl)-1,3,5-triazine-2,4,6-triamine Dihydrochloride, Monoguanylmelamine Dihydrochloride, Metformin EP Impurity B
IUPAC-namn 1-(4,6-diamino-1,3,5-triazin-2-yl)guanidine hydrochloride
LEDER N=C(NC1=NC(N)=NC(N)=N1)N.Cl.Cl
Specifikationer
Kemiskt namn eller material Guanylmelamine Dihydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C4H8N8.2ClH/c5-1(6)9-4-11-2(7)10-3(8)12-4;;/h(H8,5,6,7,8,9,10,11,12);2*1H
Formel vikt 240.05