Läs mer
Guanylurea, TRC
Produktkod. 17932095
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17932095 25 mg
17960515 250 mg
17964015 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17932095 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG84449525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 141-83-3
Molekylformel C2 H6 N4 O
Molekylvikt (g/mol) 102.1
Synonym Urea, (aminoiminomethyl)- (9CI), Urea, amidino- (6CI,7CI,8CI), N-(Aminoiminomethyl)urea, 1-Amidinourea, 1-Carbamoylguanidine, Amidinourea, Dicyandiamidine, Guanidine, (aminocarbonyl)-, Guanidinecarboxamide, Guanylurea, N-Amidinourea, N-Carbamoylguanidine, Urea, N-(aminoiminomethyl)-
IUPAC-namn carbamimidoylurea
LEDER NC(=N)NC(=O)N
Specifikationer
Kemiskt namn eller material Guanylurea
Kvantitet 25 mg
InChI formel InChI=1S/C2H6N4O/c3-1(4)6-2(5)7/h(H6,3,4,5,6,7)
Formel vikt 102.0542