Learn More
Cytiva GV050 Borosilicate Glass Replacement Lower Glass Base Assembly
The Lower Glass Base Assembly is a replacement part for Whatman™ GV 050 Series Vacuum Filtration Manifolds.
Brand: Cytiva 10442201
Este artĂculo no se puede devolver.
Vea la polĂtica de devoluciones
DescripciĂłn
- Description: Replacement Part for GV 050 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Especificaciones
Autoclavable | |
Whatman GV050/2 Glass Vacuum Filter Holder | |
Non-sterile | |
GV050/2/02 | |
GV050 | |
Filter Funnel (Base Section) |
Reusable | |
Borosilicate Glass | |
Vacuum Filtration | |
1 Ea. | |
NS29 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0443453 | Certificate of Analysis | 12/02/2025 | 197440025,10442201,197445000,197440250 |
CorrecciĂłn del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
TĂtulo del producto
Cytiva GV050 Borosilicate Glass Replacement Lower Glass Base Assembly > Lower Glass Base Assembly; Model: GV050/2/02; Type: Filter Funnel (Base Section, NS29); Material: Borosilicate Glass; For Use With: Whatman GV050/2 Glass Vacuum Filter Holder
ÂżDetecta una oportunidad de mejora?Comparta una correcciĂłn de contenido