Läs mer
Tocris Bioscience™ GW 3965 hydrochloride
GREENER_CHOICE
Produktkod. 18034612
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18034612 1 mg
18003812 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18034612 Leverantör Tocris Bioscience™ Leverantörsnummer 2474/1
Orally active liver X receptor (LXR) agonist
Selective, orally active non-steroidal agonist for the liver X receptor (LXR). In cell-based reporter gene assays, acts as a full agonist of hLXRα and hLXRβ (EC50 values are 190 and 30 nM respectively). Reduces angiotensin II-mediated increases in blood pressure; up-regulates ABCA1 gene expression and raises circulating HDL levels. Displays potent antiatherogenic activity in mouse models of atherosclerosis.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 405911-17-3
Molekylformel C33H32Cl2F3NO3
Specifikationer
Kemiskt namn eller material GW 3965 hydrochloride
Kvantitet 1 mg
Formel vikt 618.51
Renhet >98%
Molekylvikt (g/mol) 618.518