Läs mer
GW 3965 Hydrochloride, TRC
Produktkod. 30407369
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30407369 25 mg
1 options
Produktkod. 30407369 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG93098525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material GW 3965 Hydrochloride
CAS 405911-17-3
Molekylformel C33H31ClF3NO3 •HCl
Kvantitet 25 mg
Synonym GW 3965A Hydrochloride, 2-(3-(3-((2-Chloro-3-(trifluoromethyl)benzyl)(2,2-diphenylethyl)amino)propoxy)phenyl)acetic Acid Hydrochloride, 3-[3-[N-(2-Chloro-3-trifluoromethylbenzyl)(2,2-diphenylethyl)amino]propoxy]phenylacetic Acid Hydrochloride, 3-[3-[[[2-Chloro-3-(trifluoromethyl)phenyl]methyl](2,2-diphenylethyl)amino]propoxy]-benzeneacetic Acid Hydrochloride
InChI formel InChI=1S/C33H31ClF3NO3.ClH/c34-32-27(15-8-17-30(32)33(35,36)37)22-38(18-9-19-41-28-16-7-10-24(20-28)21-31(39)40)23-29(25-11-3-1-4-12-25)26-13-5-2-6-14-26;/h1-8,10-17,20,29H,9,18-19,21-23H2,(H,39,40);1H
LEDER ClC(C(C(F)(F)F)=CC=C1)=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.Cl
Molekylvikt (g/mol) 582.053646