Läs mer
Tocris Bioscience™ GW 441756
GREENER_CHOICE
Produktkod. 30001315
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001315 50 mg
18035282 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001315 Leverantör Tocris Bioscience™ Leverantörsnummer 2238/50
Potent, selective TrkA inhibitor
Potent, selective inhibitor of the NGF receptor tyrosine kinase A (TrkA) (IC50 = 2 nM). Displays > 100-fold selectivity over a range of other kinases.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 504433-23-2
Molekylformel C17H13N3O
Specifikationer
Kemiskt namn eller material GW 441756
Kvantitet 50 mg
Formel vikt 275.31
Renhet >99%
Molekylvikt (g/mol) 275.311