Läs mer
Tocris Bioscience™ GW 803430
GREENER_CHOICE
Produktkod. 18074872
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18074872 10 mg
18073842 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18074872 Leverantör Tocris Bioscience™ Leverantörsnummer 4242/10
MCH1 antagonist
Selective melanin-concentrating hormone receptor 1 (MCH1) antagonist (IC50 = 9.3 nM). Displays antiobesity and antidepressant-like effects in rats and mice. Orally active.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 515141-51-2
Molekylformel C25H24ClN3O3S
Specifikationer
Kemiskt namn eller material GW 803430
Kvantitet 10 mg
Formel vikt 481.99
Renhet >99%
Molekylvikt (g/mol) 481.995