Läs mer
GW4869, MedChemExpress
Produktkod. 16410218
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16410218 5 mg
16487748 10 mg
16430316 2 mg
16413727 50 mg
16403646 25 mg
16477606 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16410218 Leverantör Medchem Express Leverantörsnummer HY19363
MedChemExpress GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. GW4869 is an inhibitor of exosome biogenesis/release.
GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. GW4869 is an inhibitor of exosome biogenesis/release.
Specifikationer
Kemiskt namn eller material GW4869
Fysisk form Solid
CAS 6823-69-4
Färg Light Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C30H30Cl2N6O2
För användning med (applikation) COVID-19-immunoregulation
Löslighetsinformation DMSO : 0.1 mg/mL (0.17 mM; Need ultrasonic) ∣0.1 M HCL : < 1 mg/mL (ultrasonic;adjust pH to 2 with HCl) (insoluble) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
LEDER O=C(NC1=CC=C(C2=NCCN2)C=C1)/C=C/C3=CC=C(/C=C/C(NC4=CC=C(C5=NCCN5)C=C4)=O)C=C3.[H]Cl.[H]Cl
Molekylvikt (g/mol) 577.5
Formel vikt 577.5
Procent renhet 95.57%