Läs mer
Tocris Bioscience™ GX 201
Beskrivning
GX 201 is a potent and selective Nav1.7 blocker (IC50 = 3.2 nM). Exhibits 10x selectivity for Nav1.7 over Nav1.1, Nav1.2, and Nav1.6 channels. Displays prolonged residency time on the Nav1.7 channel. Chronic dosing increases compound potency ∽10-fold and provides efficacy that persists after the compound has cleared from plasma. Suppresses neuropathic pain in a mouse diabetic neuropathy model. Analgesic.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-[[1-[[2-Chloro-5-(trifluoromethyl)phenyl]methyl]-4-piperidinyl]methoxy]-5-cyclopropyl-2-fluoro-N-(methylsulfonyl)benzamide |
| CAS | 1788071-27-1 |
| Kvantitet | 10 mg |
| Mål | Voltage-gated Sodium Channel Blockers |
| Molekylformel | C25H27ClF4N2O4S |
| Renhet | 0.98 |