Learn More
GYPA Mouse anti-Human, Clone: SPM599, Abnova™
Mouse monoclonal antibody raised against full length recombinant human GYPA.
Brand: Abnova MAB13059.100ug
Additional Details : Weight : 0.02000kg
Description
Glycophorins A (GYPA) and B (GYPB) are major sialoglycoproteins of the human erythrocyte membrane which bear the antigenic determinants for the MN and Ss blood groups. In addition to the M or N and S or s antigens that commonly occur in all populations, about 40 related variant phenotypes have been identified. These variants include all the variants of the Miltenberger complex and several isoforms of Sta, as well as Dantu, Sat, He, Mg, and deletion variants Ena, S-s-U- and Mk. Most of the variants are the result of gene recombinations between GYPA and GYPB. [provided by RefSeq]
Specifications
GYPA | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
CD235a/GPA/GPErik/GPSAT/GpMiIII/HGpMiIII/HGpMiV/HGpMiX/HGpMiXI/HGpSta(C)/MN/MNS | |
Mouse | |
Protein A/G purification | |
RUO | |
2993 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
SPM599 | |
Mouse monoclonal antibody raised against full length recombinant human GYPA. | |
In 10mM PBS. | |
GYPA | |
Recombinant protein corresponding to full length of human GYPA. | |
100 μg | |
Primary | |
Human | |
Liquid |