Beskrivning
Positive Samples: Human serum Immunogen sequence: MYGKIIFVLL LSEIVSISAL STTEVAMHTS TSSSVTKSYI SSQTNGETGQ LVHRFTVPAP VVIILIILCV MAGIIGTILL ISYTIRRLIK A.
This protein is a minor sialoglycoprotein in erythrocyte membranes.
Specifikationer
Specifikationer
| Antigen | GYPB |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1.64 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gen | GYPB |
| Genaccessionsnr. | P06028 |
| Gene Alias | CD235b; glycophorin B (MNS blood group); glycophorin B/glycophorin A fusion protein; glycophorin Hop; glycophorin-B; GPB; GYP; GYPB; GYPB/GYPA fusion; hybrid glycophorin Kip; MNS; PAS-3; Sialoglycoprotein delta; SS; Ss blood group; SS-active sialoglycoprotein |
| Gensymboler | GYPB |
| Visa mer |