Läs mer
Invitrogen™ H-cadherin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581934
Beskrivning
Immunogen sequence: GTGTLLITLE DVNDNAPFIY PTVAEVCDDA KNLSVVILGA SDKDLHPNTD PFKFEIHKQA VPDKVWKISK INNTHALVSL LQNLNKANYN LPIMVTDSGK PPMTNITDLR VQVCSCRNSK VDCNAAGALR FSLPSVLLLS LFSLA Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.
The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifikationer
| H-cadherin | |
| Polyclonal | |
| Unconjugated | |
| CDH13 | |
| 4932416G01Rik; CAD13; cadherin 13; cadherin 13, H-cadherin (heart); cadherin-13; CD39 antigen-like 3; CD39L3; CD39-like 3; CDH13; CDHH; Cdht; ecto-apyrase 3; ecto-ATPase 3; ecto-ATPDase 3; HB6; H-cadherin; H-cadherin (heart); heart cadherin; heart-cadherin; NTPDase 3; NTPDase-3; P105; T cad; T Cadherin; Tcad; T-cad; T-cadherin; Truncated cadherin; truncated-cadherin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1012 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55290 | |
| CDH13 | |
| Recombinant protein corresponding to Human CDH13. Recombinant protein control fragment (Product #RP-94788). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.