Läs mer
H-Lys(Me)2-OH Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | H-Lys(Me)2-OH Hydrochloride |
| CAS | 79416-87-8 |
| Molekylformel | C8H18N2O2 . HCl |
| Kvantitet | 25 mg |
| Synonym | L-Lysine, N6,N6-dimethyl-, hydrochloride (1:1) (ACI), L-Lysine, N6,N6-dimethyl-, monohydrochloride (9CI), N6,N6-Dimethyl-L-lysine monohydrochloride, (2S)-2-Amino-6-(dimethylamino)hexanoic acid monohydrochloride, (S)-6-(Dimethylamino)-2-aminohexanoic acid monohydrochloride, Dimethyllysine hydrochloride, N,N-Dimethyllysine monohydrochloride, Nε,Nε-Dimethyl-L-lysine monohydrochloride, Nε,Nε-Dimethyllysine hydrochloride, Nε-Dimethyl-L-lysine hydrochloride, Nε-Dimethyllysine hydrochloride |
| InChI formel | InChI=1S/C8H18N2O2.ClH/c1-10(2)6-4-3-5-7(9)8(11)12;/h7H,3-6,9H2,1-2H3,(H,11,12);1H/t7-;/m0./s1 |
| LEDER | Cl.CN(C)CCCC[C@H](N)C(=O)O |
| IUPAC-namn | (2S)-2-amino-6-(dimethylamino)hexanoic acid;hydrochloride |
| Molekylvikt (g/mol) | 210.7 |
| Formel vikt | 210.1135 |