Läs mer
H-Lys(z)-OtBu Hydrochloride, TRC
Produktkod. 30464689
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464689 250 mg
30475834 100 mg
30480970 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464689 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL488850250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5978-22-3
Molekylformel C18H29ClN2O4
Molekylvikt (g/mol) 372.89
IUPAC-namn tert-butyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride
LEDER Cl.CC(C)(C)OC(=O)[C@@H](N)CCCCNC(=O)OCc1ccccc1
Specifikationer
Kemiskt namn eller material H-Lys(z)-OtBu Hydrochloride
InChI formel InChI=1S/C18H28N2O4.ClH/c1-18(2,3)24-16(21)15(19)11-7-8-12-20-17(22)23-13-14-9-5-4-6-10-14;/h4-6,9-10,15H,7-8,11-13,19H2,1-3H3,(H,20,22);1H/t15-;/m0./s1
Kvantitet 250 mg
Formel vikt 372.1816