Läs mer
H-Met-Met-Met-OH, TRC
Produktkod. 30465927
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30465927 10 mg
1 options
Produktkod. 30465927 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22215010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material H-Met-Met-Met-OH
CAS 14486-15-8
Molekylformel C15H29N3O4S3
Kvantitet 10 mg
Synonym L-Methionine, N-(N-L-methionyl-L-methionyl)-, Methionine, N-(N-L-methionyl-L-methionyl)-, L- (8CI), L-Methionyl-L-methionyl-L-methionine, Trimethionine
InChI formel InChI=1S/C15H29N3O4S3/c1-23-7-4-10(16)13(19)17-11(5-8-24-2)14(20)18-12(15(21)22)6-9-25-3/h10-12H,4-9,16H2,1-3H3,(H,17,19)(H,18,20)(H,21,22)/t10-,11-,12-/m0/s1
LEDER CSCC[C@H](N)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CCSC)C(=O)O
IUPAC-namn (2S)-2-[[(2S)-2-[[(2S)-2-amino-4-methylsulfanylbutanoyl]amino]-4-methylsulfanylbutanoyl]amino]-4-methylsulfanylbutanoic acid
Molekylvikt (g/mol) 411.6
Formel vikt 411.132